Chemical Name or MaterialTungsten, plasma standard solution, Specpure™
MDL NumberMFCD00011461
Molecular Formula(NH4)2WO4 in H2O
Physical FormLiquid
Quantity500 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.