Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL
Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL
Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL
Thermo Scientific Chemicals

Tungsten, plasma standard solution, Specpure™ W 10,000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 042249.AE
also known as 042249-AE
Price (USD)
149.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
149.00
Each
Specifications
Chemical Name or MaterialTungsten plasma standard solution
MDL NumberMFCD00011461
Name NoteW 10,000μg/mL
Physical FormLiquid
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only