Tetrahydrofuran, anhydrous, 99.8+%, unstab.
Tetrahydrofuran, anhydrous, 99.8+%, unstab.
Tetrahydrofuran, anhydrous, 99.8+%, unstab.
Thermo Scientific Chemicals

Tetrahydrofuran, anhydrous, 99.8+%, unstab.

For Synthesis and Liquid Chromatography | CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 L
Catalog number 042254.K7
also known as 042254-K7
Price (USD)
228.65
Special offer
Online exclusive
Ends: 30-Jun-2026
269.00
Save 40.35 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
228.65
Special offer
Online exclusive
Ends: 30-Jun-2026
269.00
Save 40.35 (15%)
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
CommentSourced in UK and US
Water Content (Karl Fischer Titration)≤30ppm (US Sourced Material)
Residue on Evaporation≤0.001% (≤10ppm, UK Sourced Material)
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only