Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Puratronic™, 99.999% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Puratronic™, 99.999% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, Puratronic™, 99.999% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 042259.30
also known as 042259-30
Price (USD)
623.65
Online Exclusive
693.00
Save 69.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
623.65
Online Exclusive
693.00
Save 69.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7429-90-5
ChEBICHEBI:33629
Chemical Name or MaterialAluminum slug
EINECS Number231-072-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only