Dimethyl sulfoxide-d6, 100% (Isotopic), Thermo Scientific Chemicals
Dimethyl sulfoxide-d6, 100% (Isotopic), Thermo Scientific Chemicals
Dimethyl sulfoxide-d6, 100% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl sulfoxide-d6, 100% (Isotopic), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
042285.KV
also known as 042285-KV
10 Ea.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042285.KV
also known as 042285-KV
Price (USD)
-
Quantity:
10 Ea.
Request bulk or custom format
Chemical Identifiers
CAS2206-27-1
IUPAC Name(²H₃)methanesulfinyl(²H₃)methane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid / low melting solid
Assay from Suppliers CofA≥99.5% by GC
Assay from Suppliers CofA≥99.945% Isotopic enrichment by ¹H NMR
Water Content (Karl Fischer Titration)≤ 150ppm
Deuterated DMSO is a common solvent used in NMR spectroscopy. In combination with pyridine-d5, it plays an important role for solution-state 2D NMR of ball-milled plant cell wall gel. Low-energy collisions of ionized acetone-d6, DMSO-d6 and pyridine-d5 with alkanethiolate is used to prepare self-assembled monolayer surfaces of gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only