Quartz rod, fused, 2.0mm (0.079in) dia
Quartz rod, fused, 2.0mm (0.079in) dia
Quartz rod, fused, 2.0mm (0.079in) dia
Thermo Scientific Chemicals

Quartz rod, fused, 2.0mm (0.079in) dia

CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
12 in.
48 in.
Catalog number 042292.NK
also known as 042292-NK
Price (USD)
27.90
Each
Quantity:
12 in.
Request bulk or custom format
Price (USD)
27.90
Each
Specifications
Boiling Point2230°C
CAS14808-60-7
ChEBICHEBI:30563
Chemical Name or MaterialQuartz rod
EINECS Number238-878-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only