2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol
2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol
2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol
Thermo Scientific Chemicals

2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol

Monomer used in the preparation of polymethacrylate polymers | CAS: 688-84-6 | C12H22O2 | 198.306 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 L
20 L
Catalog number 042303.K2
also known as 042303-K2
Price (USD)
69.65
Online Exclusive
77.10
Save 7.45 (10%)
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
77.10
Save 7.45 (10%)
Each
Chemical Identifiers
CAS688-84-6
IUPAC Name2-ethylhexyl 2-methylprop-2-enoate
Molecular FormulaC12H22O2
InChI KeyWDQMWEYDKDCEHT-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤10
Stabilizer40-60ppm MEHQ
Assay from Supplier's CofA≥97.5%
Monomer used in the preparation of polymethacrylate polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only