Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.98% (metals basis)
Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.98% (metals basis)
Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.98% (metals basis)
Thermo Scientific Chemicals

Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.98% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 042325.09
also known as 042325-09
Price (USD)
59.65
Online Exclusive
66.10
Save 6.45 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
59.65
Online Exclusive
66.10
Save 6.45 (10%)
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Length2.8 - 3.5mm
Total Metal Impurities0.02% max.
Appearance (Color)Gray
FormSlug
Diameter2.8 - 3.5mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only