Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.995% (metals basis)
Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.995% (metals basis)
Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Nickel slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, Puratronic™, 99.995% (metals basis)

Form: approx. 0.22g/slug | CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 042333.09
also known as 042333-09
Price (USD)
72.65
Online Exclusive
81.00
Save 8.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
81.00
Save 8.35 (10%)
Each
Specifications
AppearanceSilver gray slug
Assay Percent Range99.995% (Metals basis)
CAS7440-02-0
ChEBICHEBI:28112
Chemical Name or MaterialNickel
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only