n-Pentane-d{12}, 98%(Isotopic)
n-Pentane-d{12}, 98%(Isotopic)
n-Pentane-d{12}, 98%(Isotopic)
Thermo Scientific Chemicals

n-Pentane-d{12}, 98%(Isotopic)

CAS: 2031-90-5 | C5H12 | 84.224 g/mol
Have Questions?
Catalog NumberQuantity
042340.03
also known as 042340-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042340.03
also known as 042340-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Boiling Point36°C
CAS2031-90-5
Chemical Name or Materialn-Pentane-d{12}
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES
Density0.731 g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only