1,1,2,2-Tetrachloroethane-d{2}, 99.5% (Isotopic)
1,1,2,2-Tetrachloroethane-d{2}, 99.5% (Isotopic)
1,1,2,2-Tetrachloroethane-d{2}, 99.5% (Isotopic)
Thermo Scientific Chemicals

1,1,2,2-Tetrachloroethane-d{2}, 99.5% (Isotopic)

CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042347.04
also known as 042347-04
Price (USD)
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS33685-54-0
IUPAC Nametetrachloro(²H₂)ethane
Molecular FormulaC2H2Cl4
InChI KeyQPFMBZIOSGYJDE-QDNHWIQGSA-N
SMILES[2H]C(Cl)(Cl)C([2H])(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Colorless
Isotopic purity≥99.5%
1,1,2,2-Tetrachloroethane-d_2 is used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only