Cobalt slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)
Cobalt slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)
Cobalt slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)
Thermo Scientific Chemicals

Cobalt slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 042353.14
also known as 042353-14
Price (USD)
104.65
Special offer
Online exclusive
Ends: 30-Jun-2026
123.00
Save 18.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
104.65
Special offer
Online exclusive
Ends: 30-Jun-2026
123.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only