Nickel Chromium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals
Nickel Chromium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals
Nickel Chromium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel Chromium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
042368.18
also known as 042368-18
50 g
Catalog number 042368.18
also known as 042368-18
Price (USD)
197.65
Special offer
Online exclusive
Ends: 30-Jun-2026
232.00
Save 34.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
197.65
Special offer
Online exclusive
Ends: 30-Jun-2026
232.00
Save 34.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeNi:Cr; 80:20 wt%
CAS11106-97-1
Chemical Name or MaterialNickel Chromium slug
Health Hazard 1H317-H351
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only