Tantalum rod, 2.8mm (0.11in) dia, 99.95% (metals basis)
Tantalum rod, 2.8mm (0.11in) dia, 99.95% (metals basis)
Tantalum rod, 2.8mm (0.11in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Tantalum rod, 2.8mm (0.11in) dia, 99.95% (metals basis)

Form: Approx. 102g/m | CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 042371.BU
also known as 042371-BU
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Quantity:
25 cm
Request bulk or custom format
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Specifications
AppearanceGray to silver rod
Assay Percent Range99.95% (Metals basis)
CAS7440-25-7
ChEBICHEBI:33348
Chemical Name or MaterialTantalum
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only