Hexanes, mixed isomers, Environmental Grade
Hexanes, mixed isomers, Environmental Grade
Hexanes, mixed isomers, Environmental Grade
Thermo Scientific Chemicals

Hexanes, mixed isomers, Environmental Grade

CAS: 92112-69-1 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 042386.K2
also known as 042386-K2
Price (USD)
54.60
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
54.60
Each
Chemical Identifiers
CAS92112-69-1
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMixure of isomers: Typically 85+% n-hexane 99.8% C₆ isomers.
Residual halide10 ng/L (GC-ECD) Note: Submicron filtered.
Residue on Evaporation2ppm
Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. It is also involved in the preparation of organolithiums compounds namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only