Titanium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Puratronic™, 99.995% (metals basis)
Titanium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Puratronic™, 99.995% (metals basis)
Titanium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Titanium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Puratronic™, 99.995% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 Slugs
25 Slugs
Catalog number 042392.QJ
also known as 042392-QJ
Price (USD)
49.10
Each
Quantity:
5 Slugs
Request bulk or custom format
Price (USD)
49.10
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-32-6
ChEBICHEBI:33341
Chemical Name or MaterialTitanium slug
EINECS Number231-142-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only