Poly(2-ethyl-2-oxazoline), M.W. 200,000
Poly(2-ethyl-2-oxazoline), M.W. 200,000
Poly(2-ethyl-2-oxazoline), M.W. 200,000
Poly(2-ethyl-2-oxazoline), M.W. 200,000
Thermo Scientific Chemicals

Poly(2-ethyl-2-oxazoline), M.W. 200,000

CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 042397.A4
also known as 042397-A4
Price (USD)
364.65
Special offer
Online exclusive
Ends: 30-Jun-2026
429.00
Save 64.35 (15%)
Each
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
364.65
Special offer
Online exclusive
Ends: 30-Jun-2026
429.00
Save 64.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS25805-17-8
Molecular Formula(C5H9NO)n
SMILESCCC(=O)N(-*)CC-*
MDL NumberMFCD00134377
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to pale yellow
Formsolid
Loss on Drying2%, max ( 1g sample, 2 hours @ 125 deg C)
Viscosity18 to 24 cSt ( Kinematic viscosity of a 10 % aq. solution at 100 deg F):
Poly(2-ethyl-2-oxazoline) shows the application in biocompatibility, avoiding accumulation in tissue, and rapid clearance from the bloodstream. It is also adapted to many different functions for both protein and small drug conjugation. It also acts as a perfect substitute for poly(vinyl alcohol) and poly(vinyl pyrrolidone). It is also used as an adhesion promoter in coatings and heat sealing. It behaves as a hydrophilic block and utilized in the synthesis of amphiphilic diblock copolymers by reacting with poly(1,3-trimethylene carbonate) (PTMC) as a hydrophobic block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only