Methyl eicosanoate, 98+%
Methyl eicosanoate, 98+%
Methyl eicosanoate, 98+%
Thermo Scientific Chemicals

Methyl eicosanoate, 98+%

CAS: 1120-28-1 | C21H42O2 | 326.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 042399.04
also known as 042399-04
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.50
Save 10.85 (14%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.50
Save 10.85 (14%)
Each
Chemical Identifiers
CAS1120-28-1
IUPAC Namemethyl icosanoate
Molecular FormulaC21H42O2
InChI KeyQGBRLVONZXHAKJ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay from Suppliers CofA≥97.5%
FormPowder or crystalline powder or crystals or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only