(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles
(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles
(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 2530-87-2 | C6H15ClO3Si | 198.718 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Catalog number 042413.K2
also known as 042413-K2
Price (USD)
221.65
Special offer
Online exclusive
Ends: 30-Jun-2026
260.00
Save 38.35 (15%)
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
221.65
Special offer
Online exclusive
Ends: 30-Jun-2026
260.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS2530-87-2
IUPAC Name(3-chloropropyl)trimethoxysilane
Molecular FormulaC6H15ClO3Si
InChI KeyOXYZDRAJMHGSMW-UHFFFAOYSA-N
SMILESCO[Si](CCCCl)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay from Suppliers CofA≥97%
(3-Chloropropyl)trimethoxysilane is used as adhesives and sealant chemicals. It is also used in the preparation of 3-iodopropyltrimethoxysilane by reacting with sodium iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only