Buffer solution, pH 5.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable
Thermo Scientific Chemicals

Buffer solution, pH 5.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 042417.K7
also known as 042417-K7
Price (USD)
77.65
Online Exclusive
86.50
Save 8.85 (10%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
86.50
Save 8.85 (10%)
Each
Specifications
Chemical Name or MaterialBuffer solution, pH 5.00 (±0.01 at 25°C)
MDL NumberMFCD00147253
Name NoteNIST Traceable
Physical FormLiquid
Quantity4 L
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only