Antimony, Oil based standard solution, Specpure™ Sb 5000μg/g, Thermo Scientific Chemicals
Antimony, Oil based standard solution, Specpure™ Sb 5000μg/g, Thermo Scientific Chemicals
Antimony, Oil based standard solution, Specpure™ Sb 5000μg/g, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony, Oil based standard solution, Specpure™ Sb 5000μg/g, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
042433.18
also known as 042433-18
50 g
Catalog number 042433.18
also known as 042433-18
Price (USD)
63.65
Online Exclusive
70.90
Save 7.25 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.90
Save 7.25 (10%)
Each
Specifications
Chemical Name or MaterialAntimony, Oil based standard solution
MDL NumberMFCD00134030
Name NoteSb 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only