Calcium, Oil based standard solution, Specpure™ Ca 5000μg/g
Thermo Scientific Chemicals

Calcium, Oil based standard solution, Specpure™ Ca 5000μg/g

 
Have Questions?
Catalog NumberQuantity
042437.18
also known as 042437-18
50 g
Catalog number 042437.18
also known as 042437-18
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Specifications
Chemical Name or MaterialCalcium, Oil based standard solution
MDL NumberMFCD00085314
Name NoteCa 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only