Molybdenum, Oil based standard solution, Specpure™ Mo 5000μg/g
Thermo Scientific Chemicals

Molybdenum, Oil based standard solution, Specpure™ Mo 5000μg/g

 
Have Questions?
Catalog NumberQuantity
042445.18
also known as 042445-18
50 g
Catalog number 042445.18
also known as 042445-18
Price (USD)
78.10
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
78.10
Each
Specifications
Chemical Name or MaterialMolybdenum, Oil based standard solution
MDL NumberMFCD00003465
Name NoteMo 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only