Nickel, Oil based standard solution, Specpure™ Ni 5000μg/g
Nickel, Oil based standard solution, Specpure™ Ni 5000μg/g
Nickel, Oil based standard solution, Specpure™ Ni 5000μg/g
Thermo Scientific Chemicals

Nickel, Oil based standard solution, Specpure™ Ni 5000μg/g

 
Have Questions?
Catalog NumberQuantity
042446.18
also known as 042446-18
50 g
Catalog number 042446.18
also known as 042446-18
Price (USD)
84.65
Online Exclusive
93.70
Save 9.05 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
84.65
Online Exclusive
93.70
Save 9.05 (10%)
Each
Specifications
Chemical Name or MaterialNickel, Oil based standard solution
Health Hazard 1H350
Health Hazard 2GHS H Statement
H350
May cause cancer.
Health Hazard 3P201-P202-P281-P308+P313-P501c
MDL NumberMFCD00011137
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only