Phosphorus, Oil based standard solution, Specpure™ P 5000μg/g
Thermo Scientific Chemicals

Phosphorus, Oil based standard solution, Specpure™ P 5000μg/g

 
Have Questions?
Catalog NumberQuantity
042447.18
also known as 042447-18
50 g
Catalog number 042447.18
also known as 042447-18
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.10
Save 13.45 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.10
Save 13.45 (15%)
Each
Specifications
Chemical Name or MaterialPhosphorus, Oil based standard solution
MDL NumberMFCD00133771
Name NoteP 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only