Sodium, Oil based standard solution, Specpure™ Na 5000μg/g
Thermo Scientific Chemicals

Sodium, Oil based standard solution, Specpure™ Na 5000μg/g

 
Have Questions?
Catalog NumberQuantity
042449.18
also known as 042449-18
50 g
Catalog number 042449.18
also known as 042449-18
Price (USD)
84.65
Online Exclusive
94.20
Save 9.55 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
84.65
Online Exclusive
94.20
Save 9.55 (10%)
Each
Specifications
Chemical Name or MaterialSodium, Oil based standard solution
MDL NumberMFCD00085307
Name NoteNa 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only