Titanium, Oil based standard solution, Specpure™ Ti 5000μg/g
Titanium, Oil based standard solution, Specpure™ Ti 5000μg/g
Titanium, Oil based standard solution, Specpure™ Ti 5000μg/g
Thermo Scientific Chemicals

Titanium, Oil based standard solution, Specpure™ Ti 5000μg/g

 
Have Questions?
Catalog NumberQuantity
042451.18
also known as 042451-18
50 g
Catalog number 042451.18
also known as 042451-18
Price (USD)
77.40
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
77.40
Each
Specifications
Chemical Name or MaterialTitanium, Oil based standard solution
MDL NumberMFCD00011264
Name NoteTi 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only