Zinc, Oil based standard solution, Specpure™ Zn 5000μg/g
Thermo Scientific Chemicals

Zinc, Oil based standard solution, Specpure™ Zn 5000μg/g

 
Have Questions?
Catalog NumberQuantity
042453.18
also known as 042453-18
50 g
Catalog number 042453.18
also known as 042453-18
Price (USD)
94.20
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
94.20
Each
Specifications
Chemical Name or MaterialZinc, Oil based standard solution
MDL NumberMFCD00011291
Name NoteZn 5000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only