Methoxytrimethylsilane, 97+%
Methoxytrimethylsilane, 97+%
Methoxytrimethylsilane, 97+%
Thermo Scientific Chemicals

Methoxytrimethylsilane, 97+%

CAS: 1825-61-2 | C4H12OSi | 104.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 042464.30
also known as 042464-30
Price (USD)
264.65
Online Exclusive
294.00
Save 29.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
264.65
Online Exclusive
294.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS1825-61-2
IUPAC Namemethoxytrimethylsilane
Molecular FormulaC4H12OSi
InChI KeyPOPACFLNWGUDSR-UHFFFAOYSA-N
SMILESCO[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.0%
Refractive Index1.3655-1.3705 @ 20°C
FormLiquid
Methoxytrimethylsilane plays an important role in the synthesis of organic compounds and used as water disgusting. It is used in the preparation of alfa-lithioalkoxysilanes by treating with tert-butyllithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only