Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000
Thermo Scientific Chemicals

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Catalog number 042502.K2
also known as 042502-K2
Price (USD)
116.65
Special offer
Online exclusive
Ends: 30-Jun-2026
137.00
Save 20.35 (15%)
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
116.65
Special offer
Online exclusive
Ends: 30-Jun-2026
137.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
FormViscous liquid
Appearance (Color)Colorless
ViscosityViscosity: 180-220 cSt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only