Chromate, Ion chromatography standard solution, Specpure™ CrO4∣-2 1000μg/mL
Thermo Scientific Chemicals

Chromate, Ion chromatography standard solution, Specpure™ CrO4∣-2 1000μg/mL

K2CrO4 in H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 042564.AP
also known as 042564-AP
Price (USD)
211.65
Special offer
Online exclusive
Ends: 30-Jun-2026
249.00
Save 37.35 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
211.65
Special offer
Online exclusive
Ends: 30-Jun-2026
249.00
Save 37.35 (15%)
Each
Specifications
Chemical Name or MaterialChromate
EINECS Number232-140-5
Health Hazard 1H340-H350i
Health Hazard 3P201-P202-P281-P308+P313-P501c
MDL NumberMFCD00011368
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only