Formate, Ion chromatography standard solution, Specpure™ HCO{2}∣- 1000μg/mL
Thermo Scientific Chemicals

Formate, Ion chromatography standard solution, Specpure™ HCO{2}∣- 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 042566.AE
also known as 042566-AE
Price (USD)
114.65
Special offer
Online exclusive
Ends: 30-Jun-2026
135.00
Save 20.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 30-Jun-2026
135.00
Save 20.35 (15%)
Each
Specifications
Chemical Name or MaterialFormate, Ion chromatography standard solution
MDL NumberMFCD00003297
Name NoteHCO2- 1000μg/mL
Physical FormLiquid
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only