Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Thermo Scientific Chemicals

Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)

CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
5 kg
Catalog number 042570.18
also known as 042570-18
Price (USD)
37.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
37.00
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Total Metal Impurities≤0.5%
pH6.5-7.5 (suspension 5% w/w)
Particle SizeOver size, air jet sieve alpine 60 micrometer: ≤10% (w/w)
View more
Column chromatographySilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only