Azelaic acid, 96%
Azelaic acid, 96%
Azelaic acid, 96%
Thermo Scientific Chemicals

Azelaic acid, 96%

CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 042587.22
also known as 042587-22
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Specifications
Beilstein1101094
Boiling Point286°C (100mmHg)
CAS123-99-9
ChEBICHEBI:48131
Chemical Name or MaterialAzelaic acid
View more
Azelaic acid acts as an anti-inflammatory compound. It is also exhibits anti acne activity. It up regulates expression of ant oxidative enzymes. Azelaic acid also inhibits proliferation of melanocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only