Fullerene powder, 98+% C{70}
Fullerene powder, 98+% C{70}
Fullerene powder, 98+% C{70}
Thermo Scientific Chemicals

Fullerene powder, 98+% C{70}

CAS: 115383-22-7 | C70 | 840.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 042601.03
also known as 042601-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS115383-22-7
IUPAC Namehexatriacontacyclo[11.3.54.0¹⁷,²².0²,²³.0¹⁴,¹⁸.0¹⁹,⁷⁰.0²⁰,³⁷.0²¹,³⁴.0⁸,¹⁶.0¹⁰,¹⁵.0⁹,⁶³.0³,⁷.0⁴,²⁵.0⁶,⁶².0⁵,⁴⁸.0²⁴,³³.0³¹,³⁵.0²⁷,³².0²⁶,⁴⁷.0²⁸,⁴⁶.0⁴⁵,⁵⁰.0⁴⁹,⁶¹.0⁵¹,⁵⁹.0⁴⁴,⁵².0⁶⁰,⁶⁴.0¹¹,⁶⁵.0¹²,⁶⁷.0⁵⁸,⁶⁶.0⁵⁶,⁶⁸.0³⁸,⁶⁹.0³⁹,⁵⁵.0³⁶,⁴⁰.0³⁰,⁴¹.0²⁹,⁴³.0⁴²,⁵⁴.0⁵³,⁵⁷]heptaconta-1,3,5(48),6,8(16),9,11(65),12(67),13(70),14,17(22),18,20,23,25,27(32),28,30(41),31(35),33,36,38(69),39,42,44(52),45(50),46,49(61),51(59),53(57),54,56(68),58(66),60(64),62-pentatriacontaene
Molecular FormulaC70
InChI KeyATLMFJTZZPOKLC-UHFFFAOYSA-N
SMILESC12=C3C4=C5C1=C1C6=C7C8=C9C%10=C%11C%12=C%13C%14=C%15C%16=C%17C%18=C%19C%20=C%21C%22=C%19C%19=C%23C(C%11=C%19C%18=C%12%16)=C9C9=C%11C(=C2C9=C18)C1=C(C2=C%21C8=C9C(=C4C4=C9C9=C%12C(=C%17C%20=C8%12)C%15=C8C%14=C(C%12=C6C5=C4C%12=C98)C7=C%10%13)C2=C31)C%22=C%23%11
View more
SpecificationsSpecification SheetSpecification Sheet
PurityC70: guaranteed ≥98.0%
FormPowder
Fullerene powder is used in chemical research. It is extensively used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only