Dionex™ Postcolumn Ambient Reaction Accessories (Pneumatic or Pump Delivery)
Use these accessories for Postcolumn Ambient Reaction products to ensure consistent, pulse-free reagent delivery for applications with high background absorbances.
Have Questions?
Change view
Catalog Number
Description
Capacity
042631
Knitted Reaction Coil, Unpotted
750 μL
042630
Knitted Reaction Coil, Unpotted
1500 μL
036036
Packed Reaction Coil
500 μL
3 Options
Catalog number 042631
Price (CLP)
-
Description:
Knitted Reaction Coil, Unpotted
Capacity:
750 μL
Use these accessories for Thermo Scientific™ Dionex™ Postcolumn Ambient Reaction products to ensure consistent, pulse-free reagent delivery for applications with high background absorbances, such as trace-level transition metal analysis using post-column reaction with 4-(2-pyridylazo)resorcinol (PAR).
Specifications
Capacity750 μL
DescriptionKnitted Reaction Coil, Unpotted
OperationEnsures consistent, pulse-free reagent delivery for applications with high background absorbances.