Iodine monochloride, approx. 0.22N soln. in glacial acetic acid
Iodine monochloride, approx. 0.22N soln. in glacial acetic acid
Iodine monochloride, approx. 0.22N soln. in glacial acetic acid
Iodine monochloride, approx. 0.22N soln. in glacial acetic acid
Thermo Scientific Chemicals

Iodine monochloride, approx. 0.22N soln. in glacial acetic acid

For Determination of Iodine Absorption Number of Fats | CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 042647.K7
also known as 042647-K7
Price (USD)
315.65
Online Exclusive
350.00
Save 34.35 (10%)
Each
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)
315.65
Online Exclusive
350.00
Save 34.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7790-99-0
IUPAC Nameiodochlorane
Molecular FormulaClI
InChI KeyQZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILESClI
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Brown
Assay from Supplier's CofAca. 0.22N
Assay from Supplier's CofAFurther Ingredients: Acetic acid CAS 64-19-7, EEC No 200-580-7
For determination of iodine absorption number of fatsIodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture and Light Sensitive
RUO – Research Use Only