Germanium(IV) isopropoxide, 99.9% (metals basis)
Germanium(IV) isopropoxide, 99.9% (metals basis)
Germanium(IV) isopropoxide, 99.9% (metals basis)
Germanium(IV) isopropoxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Germanium(IV) isopropoxide, 99.9% (metals basis)

CAS: 21154-48-3 | C12H28GeO4 | 308.982 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 042673.03
also known as 042673-03
Price (USD)
69.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
69.00
Each
Add to cart
Specifications
Assay Percent Notes(metals basis)
CAS21154-48-3
Chemical Name or MaterialGermanium (IV) isopropoxide
ColorColorless
Density1.025 g/mL
View more
Germanium(IV) isopropoxide is used as precursor to prepare GeO2 organic silanes, which is used to study the application of optical wave guide. It is used in the synthesis of heterotactic polyactide and also involved in the macromolecule mediated synthesis of amorphous Germania.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only