Indium tin oxide, Vacuum Deposition Grade, 99.99% (metals basis)
Indium tin oxide, Vacuum Deposition Grade, 99.99% (metals basis)
Indium tin oxide, Vacuum Deposition Grade, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium tin oxide, Vacuum Deposition Grade, 99.99% (metals basis)

CAS: 50926-11-9 | InOSn | 249.527 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042677.22
also known as 042677-22
Price (USD)
670.65
Special offer
Online exclusive
Ends: 30-Jun-2026
789.00
Save 118.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
670.65
Special offer
Online exclusive
Ends: 30-Jun-2026
789.00
Save 118.35 (15%)
Each
Chemical Identifiers
CAS50926-11-9
IUPAC Namestannanone indium
Molecular FormulaInOSn
InChI KeyAMGQUBHHOARCQH-UHFFFAOYSA-N
SMILES[In].O=[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Indium tin oxide is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only