Palladium Silver foil, 0.5mm (0.02in) thick, 99.9% (metals basis excluding Pt)
Thermo Scientific Chemicals

Palladium Silver foil, 0.5mm (0.02in) thick, 99.9% (metals basis excluding Pt)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 042684.FF
also known as 042684-FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis excluding Pt)
Avg. Mol. Wt. or Mol. Wt. RangePd:Ag; 75:25 wt%
Chemical Name or MaterialPalladium Silver foil
MDL NumberMFCD02091738
Name Note0.5mm (0.02 in.) thick
View more
Palladium silver foils are used in dentistry for dental crowns and bridges. It also used in conductive films and pastes. It is used to make multilayer capacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only