Platinum Ruthenium foil, 0.05mm (0.002in) thick
Thermo Scientific Chemicals

Platinum Ruthenium foil, 0.05mm (0.002in) thick

Pt:Ru; 95.2:4.8
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 042686.FF
also known as 042686-FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
AppearanceSilver-gary foil
Chemical Name or MaterialPlatinum Ruthenium
ColorSilver to Gray
MDL NumberMFCD00798552
Molecular FormulaPt:Ru; 95.2:4.8
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only