Holmium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Holmium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Holmium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Holmium foil, 0.025mm (0.001in) thick, 99.9% (REO)

CAS: 7440-60-0 | Ho | 164.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 042687.FF
also known as 042687-FF
Price (USD)
171.65
Special offer
Online exclusive
Ends: 30-Jun-2026
202.00
Save 30.35 (15%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
171.65
Special offer
Online exclusive
Ends: 30-Jun-2026
202.00
Save 30.35 (15%)
Each
Chemical Identifiers
CAS7440-60-0
IUPAC Nameholmium
Molecular FormulaHo
InChI KeyKJZYNXUDTRRSPN-UHFFFAOYSA-N
SMILES[Ho]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in mylar under argon
RUO – Research Use Only