Tri-n-butylgermanium hydride, 97+%
Tri-n-butylgermanium hydride, 97+%
Tri-n-butylgermanium hydride, 97+%
Thermo Scientific Chemicals

Tri-n-butylgermanium hydride, 97+%

Reduction of benzylic chlorides | CAS: 998-39-0 | C12H27Ge | 243.978 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 042688.06
also known as 042688-06
Price (USD)
485.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
485.00
Each
Chemical Identifiers
CAS998-39-0
IUPAC Nametributylgermyl
Molecular FormulaC12H27Ge
InChI KeyUMRPCNGKANTZSN-UHFFFAOYSA-N
SMILESCCCC[Ge](CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear, colorless to pale yellow
FormLiquid
Assay from Supplier's CofA≥97%
Tri-n-butylgermanium hydride is used in the reduction of benzylic chlorides. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only