Potassium chloride, 0.0145M, Conductivity Standard
Potassium chloride, 0.0145M, Conductivity Standard
Potassium chloride, 0.0145M, Conductivity Standard
Thermo Scientific Chemicals

Potassium chloride, 0.0145M, Conductivity Standard

CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 042693.K2
also known as 042693-K2
Price (USD)
51.90
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
51.90
Each
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Concentration0.0145M ± 0.39%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only