n-Octyltrimethoxysilane, 97+%
n-Octyltrimethoxysilane, 97+%
n-Octyltrimethoxysilane, 97+%
Thermo Scientific Chemicals

n-Octyltrimethoxysilane, 97+%

CAS: 3069-40-7 | C11H26O3Si | 234.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042698.22
also known as 042698-22
Price (USD)
331.65
Online Exclusive
368.00
Save 36.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
331.65
Online Exclusive
368.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS3069-40-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)> 97.0%
n-Octyltrimethoxysilane is used in medicine field, electronic industry and polymer material. It is also used as surface modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only