Polystyrene latex microsphere, 4.5 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 4.5 micron, 2.5 wt% dispersion in water
Polystyrene latex microsphere, 4.5 micron, 2.5 wt% dispersion in water
Thermo Scientific Chemicals

Polystyrene latex microsphere, 4.5 micron, 2.5 wt% dispersion in water

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
042715.AA
also known as 042715-AA
5 mL
Catalog number 042715.AA
also known as 042715-AA
Price (USD)
616.65
Online exclusive
685.00
Save 68.35
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
616.65
Online exclusive
685.00
Save 68.35
Each
Specifications
CAS9003-53-6
ChEBICHEBI:27452
Chemical Name or MaterialPolystyrene latex microsphere
Concentration or Composition (by Analyte or Components)2.5 wt% dispersion in water
IUPAC Namestyrene
View more
Polystyrene latex microspheres are utilized in flow cytometry, fluorescence microscopy, and as calibration particles. It finds a use in the detection of trace amounts of antigens or antibodies in serum, urine, and cerebrospinal fluids. It is also employed as a stimulus responsive particulate emulsifier for oil-in-water emulsion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only