Silica gel 60, 0.075-0.2mm (70-200 mesh)
Thermo Scientific Chemicals

Silica gel 60, 0.075-0.2mm (70-200 mesh)

Have Questions?
Change viewbuttonViewtableView
Particle Size:
-70/200 Mesh
70/150 Mesh
Quantity:
View all
50 g
250 g
5 kg
Surface Area (Metric):
View all
470 to 570 m2/g
Catalog number 042723.30
also known as 042723-30
Price (USD)
74.65
Online Exclusive
82.70
Save 8.05 (10%)
Each
Particle Size:
-70/200 Mesh
Quantity:
250 g
Surface Area (Metric):
470 to 570 m2/g
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
82.70
Save 8.05 (10%)
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size75-200 Microns
Particle Size60 Angstroms mean pore diameter
Classic column chromatography with no applied pressure (gravity pulled)Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only