Silica gel 60, 0.062-0.105mm (150-230 mesh)
Silica gel 60, 0.062-0.105mm (150-230 mesh)
Silica gel 60, 0.062-0.105mm (150-230 mesh)
Thermo Scientific Chemicals

Silica gel 60, 0.062-0.105mm (150-230 mesh)

Column chromatography without applied pressure (gravity pulled) | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 042727.A1
Price (USD)
277.65
Online Exclusive
308.00
Save 30.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
277.65
Online Exclusive
308.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-150+230 Mesh
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only