Silica gel 60, 0.019-0.037mm (400-600 mesh)
Silica gel 60, 0.019-0.037mm (400-600 mesh)
Silica gel 60, 0.019-0.037mm (400-600 mesh)
Thermo Scientific Chemicals

Silica gel 60, 0.019-0.037mm (400-600 mesh)

AA4272918 | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
5 kg
Catalog number 042729.A7
also known as 042729-A7
Price (USD)
901.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,061.00
Save 159.35 (15%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
901.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,061.00
Save 159.35 (15%)
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale gray
Particle Size18-37 Microns
Particle SizeMean Pore Diameter: 60 Angstroms
Surface Area500-600 m²/g
FormPowder
Column chromatography with elevated pressureSilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only