Samarium cobalt, REacton™, Sm 33%
Samarium cobalt, REacton™, Sm 33%
Samarium cobalt, REacton™, Sm 33%
Thermo Scientific Chemicals

Samarium cobalt, REacton™, Sm 33%

CAS: 12017-68-4 | CoSm | 209.293 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 042732.18
also known as 042732-18
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.30
Save 13.65
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.30
Save 13.65
Each
Chemical Identifiers
CAS12017-68-4
IUPAC Namepentacobalt samarium
Molecular FormulaCo5Sm
InChI KeyNDYCBWZIOSTTHS-UHFFFAOYSA-N
SMILES[Co].[Co].[Co].[Co].[Co].[Sm]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Sm 33% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only